Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016047.3 | NP_057131.1 | 3 | 436 | ||
GenBank mRNA | AF151868.1 | - | 3 | 432 | ||
AF161523.1 | - | 3 | 330 | |||
AF184213.1 | - | 3 | 432 | |||
AF401310.1 | - | 3 | 221 | |||
AK000621.1 | - | 3 | 336 | |||
AK311118.1 | - | 2 | 223 | |||
AK311908.1 | - | 3 | 369 | |||
BC015463.1 | - | 3 | 436 | 3914184 | ||
BG200650.1 | - | 2 | 197 |