Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016062.3 | NP_057146.1 | 3, 4 | 408 | ||
NR_024525.2 | 2, 3 | 564 | ||||
NR_046109.1 | 3, 4 | 474 | ||||
GenBank mRNA | AF151886.1 | - | 3, 4 | 384 | ||
AF161467.1 | - | 3, 4 | 373 | |||
BC001733.1 | - | 3, 4 | 368 | 3353705 | ||
BC005023.1 | - | 3, 4 | 371 | 3632169 | ||
BI598300.1 | - | 2, 3 | 559 | |||
BI603322.1 | - | 3, 4 | 438 | |||
BM091095.1 | - | 2, 3 | 488 | |||
BQ955804.1 | - | 3, 4 | 340 | |||
CD674578.1 | - | 3, 4 | 392 | |||
DR155511.1 | - | 2, 3 | 605 |