Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015509.2 | NP_001015509.1 | 3 | 1832 | ||
NM_016077.4 | NP_057161.1 | 2 | 513 | |||
XM_011524887.1 | XP_011523189.1 | 2 | 367 | |||
GenBank mRNA | AF151905.1 | - | 2 | 412 | ||
AK057033.1 | - | 3 | 1661 | |||
AK098219.1 | - | 1 | 1799 | |||
AL137322.1 | - | 1 | 278 | |||
BC006807.1 | - | 2 | 323 | 3446518 | ||
BC017457.1 | - | 3 | 1644 | |||
BX647177.1 | - | 1 | 3729 |