Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278167.1 | NP_001265096.1 | 10 | 1409 | ||
NM_001278168.1 | NP_001265097.1 | 9 | 1178 | |||
NM_001278169.1 | NP_001265098.1 | 10 | 1177 | |||
NM_016082.4 | NP_057166.4 | 10 | 1261 | |||
NM_016408.3 | NP_057492.2 | 10 | 1409 | |||
XM_011528856.2 | XP_011527158.1 | 7 | 2145 | |||
XM_011528857.2 | XP_011527159.1 | 9 | 1021 | |||
XM_011528858.1 | XP_011527160.1 | 9 | 1260 | |||
XM_011528859.2 | XP_011527161.1 | 9 | 1020 | |||
XM_011528860.1 | XP_011527162.1 | 7 | 720 | |||
XM_017027876.1 | XP_016883365.1 | 9 | 1159 | |||
XM_017027877.1 | XP_016883366.1 | 4 | 473 | |||
GenBank mRNA | AF161516.1 | - | 10 | 1357 | ||
AK023992.1 | - | 10 | 1359 | |||
AK027449.1 | - | 10 | 1209 | |||
AK095644.1 | - | 1 | 709 | |||
AK222745.1 | - | 10 | 1359 | |||
AK292075.1 | - | 10 | 1127 | |||
AK295342.1 | - | 10 | 1345 | |||
AK314532.1 | - | 10 | 1359 | |||
AY462283.1 | - | 10 | 1360 | |||
AY462284.1 | - | 10 | 1360 | |||
BC001215.1 | - | 9 | 1107 | 3535564 | ||
BC050706.1 | - | 10 | 1352 | 4418659 | ||
BM792566.1 | - | 2 | 101 | |||
CX785345.1 | - | 5 | 616 | |||
KU178644.1 | - | 8 | 1016 | |||
KU178645.1 | - | 9 | 1247 |