Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001317785.1 | NP_001304714.1 | 2 | 412 | ||
NM_001317786.1 | NP_001304715.1 | 2 | 262 | |||
NM_001317787.1 | NP_001304716.1 | 2 | 259 | |||
NM_001317788.1 | NP_001304717.1 | 2 | 259 | |||
NM_001317789.1 | NP_001304718.1 | 1 | 67 | |||
NM_016086.2 | NP_057170.1 | 2 | 409 | |||
NR_134486.1 | 2 | 259 | ||||
NR_134487.1 | 1 | 67 | ||||
XM_011516292.2 | XP_011514594.1 | 2 | 255 | |||
XM_011516293.2 | XP_011514595.1 | 2 | 255 | |||
XM_011516300.1 | XP_011514602.1 | 2 | 211 | |||
XM_017012296.1 | XP_016867785.1 | 2 | 255 | |||
XM_017012297.1 | XP_016867786.1 | 2 | 258 | |||
XM_017012298.1 | XP_016867787.1 | 2 | 218 | |||
XM_017012299.1 | XP_016867788.1 | 2 | 255 | |||
XM_017012300.1 | XP_016867789.1 | 2 | 255 | |||
XM_017012301.1 | XP_016867790.1 | 2 | 255 | |||
XM_017012302.1 | XP_016867791.1 | 2 | 258 | |||
GenBank mRNA | AF069762.1 | - | 2 | 405 | ||
AK308828.1 | - | 2 | 214 | |||
AK308912.1 | - | 2 | 236 | |||
AL533027.3 | - | 2 | 211 | |||
AY927581.1 | - | LIT3580 AY927581 |
1 | 48 | ||
BC024035.2 | - | 2 | 372 | 2988900 | ||
BI772699.1 | - | 2 | 262 | |||
BM126964.1 | - | 2 | 143 | |||
BM824947.1 | - | 2 | 383 | |||
BU859937.1 | - | 2 | 244 | |||
BX537896.1 | - | 2 | 239 |