Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270879.1 | NP_001257808.1 | 1 | 186 | ||
NM_016098.3 | NP_057182.1 | 1 | 186 | |||
NR_073093.1 | 1 | 186 | ||||
GenBank mRNA | AF125101.1 | - | 1 | 171 | ||
AK310934.1 | - | 1 | 113 | |||
AK312201.1 | - | 1 | 110 | |||
AL556427.3 | - | 1 | 105 | |||
AL561589.3 | - | 1 | 74 | |||
BC000810.1 | - | 1 | 96 | 3452973 | ||
BF665465.1 | - | 1 | 45 | |||
BG828174.1 | - | 1 | 144 | |||
DC402302.1 | - | 1 | 186 |