Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135212.1 | NP_001128684.1 | 2 | 426 | ||
NM_181342.2 | NP_851939.1 | 2 | 426 | |||
XM_005246638.4 | XP_005246695.1 | 2 | 1676 | |||
GenBank mRNA | AF092137.1 | - | 2 | 381 | ||
AF100751.1 | - | 2 | 392 | |||
AK292145.1 | - | 2 | 351 | |||
AK299215.1 | - | 2 | 351 | |||
AY353086.1 | - | 2 | 297 | |||
AY359015.1 | - | 2 | 319 | |||
BC009711.1 | - | 2 | 348 | 3891173 | ||
BT007122.1 | - | 2 | 297 | |||
DA020387.1 | - | 2 | 426 |