Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243351.1 | NP_001230280.1 | 7 | 728 | ||
NM_016118.4 | NP_057202.3 | 7 | 728 | |||
XM_005250011.2 | XP_005250068.1 | 7 | 793 | |||
XM_017012304.1 | XP_016867793.1 | 7 | 787 | |||
XM_017012305.1 | XP_016867794.1 | 7 | 787 | |||
XM_017012306.1 | XP_016867795.1 | 7 | 809 | |||
XM_017012307.1 | XP_016867796.1 | 7 | 790 | |||
XM_017012308.1 | XP_016867797.1 | 7 | 787 | |||
XM_017012309.1 | XP_016867798.1 | 7 | 793 | |||
XM_017012310.1 | XP_016867799.1 | 8 | 890 | |||
XM_017012311.1 | XP_016867800.1 | 8 | 794 | |||
XM_017012312.1 | XP_016867801.1 | 8 | 763 | |||
XM_017012313.1 | XP_016867802.1 | 8 | 890 | |||
XM_017012314.1 | XP_016867803.1 | 8 | 766 | |||
GenBank mRNA | AF155099.1 | - | 7 | 660 | ||
AF300717.1 | - | 7 | 717 | |||
AF459743.1 | - | 7 | 676 | |||
AY129295.1 | - | 7 | 676 | |||
BC046354.2 | - | 6 | 644 | 6070128 |