Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316935.1 | NP_001303864.1 | 6 | 429 | ||
NM_016126.3 | NP_057210.2 | 6 | 626 | |||
NR_133632.1 | 4 | 450 | ||||
NR_133633.1 | 7 | 724 | ||||
GenBank mRNA | AF100747.1 | - | 6 | 398 | ||
AF416725.1 | - | 5 | 353 | |||
AF417114.1 | - | 5 | 353 | |||
AK225916.1 | - | 5 | 2513 | |||
AW024033.1 | - | 3 | 200 | |||
BC005245.1 | - | 6 | 418 | 3838843 | ||
HY179788.1 | - | 6 | 430 | |||
L32869.1 | - | 4 | 201 | |||
X79302.1 | - | 4 | 201 |