Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016200.4 | NP_057284.1 | 3 | 361 | ||
GenBank mRNA | AF182294.1 | - | 3 | 251 | ||
AK090561.1 | - | 1 | 1568 | |||
AK311936.1 | - | 3 | 277 | |||
BC002742.2 | - | 3 | 207 | 3626875 | ||
BC022440.1 | - | 3 | 231 | 4283482 | ||
DA112078.1 | - | 3 | 230 | |||
DA318705.1 | - | 3 | 328 | |||
DA478457.1 | - | 3 | 229 | |||
DB275674.1 | - | 3 | 362 |