Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016200.4 | NP_057284.1 | 2 | 250 | ||
GenBank mRNA | AF182294.1 | - | 2 | 140 | ||
AK311936.1 | - | 2 | 166 | |||
BC002742.2 | - | 2 | 96 | 3626875 | ||
BC022440.1 | - | 2 | 120 | 4283482 | ||
DA112078.1 | - | 2 | 118 | |||
DA318705.1 | - | 2 | 217 | |||
DA478457.1 | - | 2 | 118 | |||
DB275674.1 | - | 2 | 251 |