Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318524.1 | NP_001305453.1 | 3 | 340 | ||
NM_001318525.1 | NP_001305454.1 | 3 | 340 | |||
NM_001318526.1 | NP_001305455.1 | 3 | 405 | |||
NM_001318527.1 | NP_001305456.1 | 3 | 656 | |||
NM_001318528.1 | NP_001305457.1 | 3 | 317 | |||
NM_001318529.1 | NP_001305458.1 | 3 | 656 | |||
NM_001318530.1 | NP_001305459.1 | 3 | 317 | |||
NM_001318532.1 | NP_001305461.1 | 3 | 656 | |||
NM_016209.4 | NP_057293.1 | 3 | 340 | |||
NR_134670.1 | 4 | 527 | ||||
NR_134671.1 | 4 | 456 | ||||
XM_017023270.1 | XP_016878759.1 | 3 | 387 | |||
GenBank mRNA | AF089106.1 | - | 3 | 333 | ||
AF161524.1 | - | 3 | 295 | |||
AK126779.1 | - | 3 | 299 | |||
AK307253.1 | - | 4 | 456 | |||
AK310805.1 | - | 3 | 611 | |||
AK311885.1 | - | 3 | 309 | |||
AX771012.1 | - | 3 | 333 | |||
AX771013.1 | - | 3 | 295 | |||
BC011369.1 | - | 3 | 287 | 3868798 | ||
BC018024.1 | - | 3 | 294 | 4287920 | ||
BC105809.1 | - | 3 | 306 | 6745065 | ||
BI754306.1 | - | 4 | 469 | |||
BX340702.2 | - | 3 | 602 | |||
BX384775.2 | - | 3 | 611 | |||
BX419849.2 | - | 3 | 285 | |||
DA209193.1 | - | 3 | 317 | |||
DA293122.1 | - | 3 | 340 | |||
DA295904.1 | - | 3 | 276 | |||
DA833474.1 | - | 3 | 279 |