Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRAPPC2L SIRNA Assays ›
Gene Symbol:
TRAPPC2L
Gene Name:
trafficking protein particle complex 2-like
Gene Aliases:
HSPC176
Chromosome Location:
Chr.16: 88856220 - 88862686 on Build GRCh38
UniGene ID:
Hs.461722
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28535
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRAPPC2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318524.1
NP_001305453.1
3, 4
366
NM_001318525.1
NP_001305454.1
3, 4
366
NM_001318526.1
NP_001305455.1
3, 4
431
NM_001318527.1
NP_001305456.1
3, 4
682
NM_001318528.1
NP_001305457.1
3, 4
343
NM_001318529.1
NP_001305458.1
3, 4
682
NM_001318530.1
NP_001305459.1
3, 4
343
NM_001318532.1
NP_001305461.1
3, 4
682
NM_016209.4
NP_057293.1
3, 4
366
NR_134670.1
4, 5
553
NR_134671.1
4, 5
482
XM_017023270.1
XP_016878759.1
3, 4
413
GenBank mRNA
AF089106.1
-
3, 4
359
AF161524.1
-
3, 4
321
AK126779.1
-
3, 4
325
AK307253.1
-
4, 5
482
AK310805.1
-
3, 4
637
AK311885.1
-
3, 4
335
AX771012.1
-
3, 4
359
AX771013.1
-
3, 4
321
BC011369.1
-
3, 4
313
3868798
BC018024.1
-
3, 4
320
4287920
BC105809.1
-
3, 4
332
6745065
BG283878.1
-
3, 4
366
BI754306.1
-
4, 5
495
BX340702.2
-
3, 4
628
BX384775.2
-
3, 4
637
BX419849.2
-
3, 4
311
DA209193.1
-
3, 4
343
DA293122.1
-
3, 4
366
DA295904.1
-
3, 4
302
DA833474.1
-
3, 4
305
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4h6qx:80/100.66.76.46:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline