Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016217.2 | NP_057301.1 | 2 | 1562 | ||
GenBank mRNA | AB033492.1 | - | 2 | 1562 | ||
BC070068.1 | - | 2 | 1570 | 30340910 |