Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016231.4 | NP_057315.3 | 8, 9 | 1458 | ||
GenBank mRNA | AF180819.1 | - | 8, 9 | 1456 | ||
AF197898.1 | - | 8, 9 | 1458 | |||
AK024686.1 | - | 8, 9 | 1090 | |||
AK315315.1 | - | 8, 9 | 1473 | |||
BC064663.1 | - | 8, 9 | 1970 | 6527673 |