Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159694.1 | NP_001153166.1 | 6, 7 | 700 | ||
NM_016242.3 | NP_057326.2 | 7, 8 | 739 | |||
XM_011532024.2 | XP_011530326.1 | 7, 8 | 698 | |||
GenBank mRNA | AB034695.1 | - | 7, 8 | 638 | ||
AF205940.1 | - | 7, 8 | 668 | |||
AK026875.1 | - | 7, 8 | 665 | |||
AK222741.1 | - | 7, 8 | 641 | |||
AK304568.1 | - | 6, 7 | 610 | |||
AK313504.1 | - | 7, 8 | 668 | |||
AY039241.1 | - | 6, 7 | 439 |