Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323565.1 | NP_001310494.1 | 4 | 717 | ||
NM_001323566.1 | NP_001310495.1 | 3 | 657 | |||
NM_001323567.1 | NP_001310496.1 | 3 | 253 | |||
NM_001801.2 | NP_001792.2 | 3 | 660 | |||
NR_136618.1 | 3 | 660 | ||||
NR_136619.1 | 3 | 660 | ||||
NR_136620.1 | 3 | 620 | ||||
NR_136621.1 | 3 | 253 | ||||
GenBank mRNA | AK130357.1 | - | 3 | 622 | ||
AK314231.1 | - | 3 | 610 | |||
AL534761.3 | - | 3 | 659 | |||
BC024241.2 | - | 3 | 610 | 4797463 | ||
BG197197.1 | - | 2 | 137 | |||
BG720517.1 | - | 3 | 258 | |||
BG897741.1 | - | 3 | 305 | |||
CK005322.1 | - | 1 | 23 | |||
CR536540.1 | - | 3 | 349 | |||
D85777.1 | - | 3 | 578 | |||
Z23010.1 | - | 3 | 349 | |||
Z31357.1 | - | 3 | 603 |