Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278666.1 | NP_001265595.1 | 2 | 268 | ||
NM_001278667.1 | NP_001265596.1 | 2 | 440 | |||
NM_001278668.1 | NP_001265597.1 | 2 | 267 | |||
NM_016277.4 | NP_057361.3 | 2 | 737 | |||
NM_183227.2 | NP_899050.1 | 2 | 291 | |||
NR_103822.1 | 2 | 268 | ||||
GenBank mRNA | AB025427.1 | - | 1 | 83 | ||
AB034244.1 | - | 2 | 236 | |||
AF161486.1 | - | 2 | 233 | |||
AF498951.1 | - | 1 | 83 | |||
AK311123.1 | - | 2 | 257 | |||
AK313796.1 | - | 2 | 288 | |||
AU280196.1 | - | 2 | 267 | |||
AY585189.1 | - | 1 | 83 | |||
BC015021.1 | - | 2 | 703 | 3910708 | ||
CR749371.1 | - | 3 | 471 | |||
DA021294.1 | - | 2 | 291 | |||
DA997359.1 | - | 2 | 436 |