Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016306.5 | NP_057390.1 | 5 | 812 | ||
GenBank mRNA | AB028859.2 | - | 5 | 755 | ||
AF228505.1 | - | 5 | 681 | |||
AF277317.1 | - | 5 | 772 | |||
AJ250137.1 | - | 5 | 610 | |||
AK075300.1 | - | 5 | 769 | |||
AK075430.1 | - | 5 | 788 | |||
AK124289.1 | - | 1 | 3208 | |||
AK301292.1 | - | 3 | 471 | |||
AY359043.1 | - | 5 | 748 | |||
BC001144.1 | - | 5 | 750 | 3534756 | ||
BC046500.1 | - | 6 | 1519 | |||
BT007063.1 | - | 5 | 590 | |||
CR457096.1 | - | 5 | 590 |