Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016332.2 | NP_057416.1 | 2 | 322 | ||
GenBank mRNA | AF161388.1 | - | 2 | 279 | ||
AF166124.1 | - | 2 | 281 | |||
AF187272.1 | - | 2 | 256 | |||
AK026128.1 | - | 2 | 293 | |||
BC003127.1 | - | 2 | 218 | 2905838 |