Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271880.1 | NP_001258809.1 | 3, 4 | 399 | ||
NM_016438.3 | NP_057522.1 | 2, 3 | 483 | |||
XM_006721946.2 | XP_006722009.1 | 3, 4 | 377 | |||
XM_006721947.3 | XP_006722010.1 | 3, 4 | 787 | |||
XM_006721948.3 | XP_006722011.1 | 3, 4 | 481 | |||
XM_011524891.2 | XP_011523193.1 | 3, 4 | 881 | |||
XM_017024742.1 | XP_016880231.1 | 3, 4 | 827 | |||
XM_017024743.1 | XP_016880232.1 | 3, 4 | 919 | |||
GenBank mRNA | AB038021.1 | - | 2, 3 | 310 | ||
AB038022.1 | - | 2, 3 | 310 | |||
BC020667.1 | - | 2, 3 | 474 | 4724613 | ||
BG573492.1 | - | 2, 3 | 475 | |||
BM503772.1 | - | 2, 3 | 486 |