Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271880.1 | NP_001258809.1 | 3 | 336 | ||
NM_016438.3 | NP_057522.1 | 2 | 420 | |||
XM_006721946.2 | XP_006722009.1 | 3 | 314 | |||
XM_006721947.3 | XP_006722010.1 | 3 | 724 | |||
XM_006721948.3 | XP_006722011.1 | 3 | 418 | |||
XM_011524891.2 | XP_011523193.1 | 3 | 818 | |||
XM_017024742.1 | XP_016880231.1 | 3 | 764 | |||
XM_017024743.1 | XP_016880232.1 | 3 | 856 | |||
GenBank mRNA | AB038021.1 | - | 2 | 247 | ||
AB038022.1 | - | 2 | 247 | |||
BC020667.1 | - | 2 | 411 | 4724613 | ||
BG573492.1 | - | 2 | 412 | |||
BM503772.1 | - | 2 | 423 |