Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042589.2 | NP_001036054.1 | 7 | 1173 | ||
NM_001042590.2 | NP_001036055.1 | 6 | 1457 | |||
NM_016446.3 | NP_057530.2 | 7 | 1245 | |||
XM_005251483.4 | XP_005251540.3 | 6 | 1427 | |||
XM_011517901.2 | XP_011516203.1 | 6 | 1427 | |||
XM_011517902.2 | XP_011516204.1 | 6 | 1427 | |||
XM_011517903.2 | XP_011516205.1 | 6 | 1427 | |||
XM_011517904.2 | XP_011516206.1 | 6 | 1628 | |||
XM_011517905.1 | XP_011516207.1 | 6 | 1427 | |||
XM_011517908.2 | XP_011516210.1 | 5 | 880 | |||
XM_011517910.2 | XP_011516212.1 | 5 | 758 | |||
XM_011517911.2 | XP_011516213.1 | 7 | 1173 | |||
XM_011517912.2 | XP_011516214.1 | 6 | 1019 | |||
XM_011517913.2 | XP_011516215.1 | 6 | 1234 | |||
XM_011517914.2 | XP_011516216.1 | 4 | 679 | |||
XM_011517915.2 | XP_011516217.1 | 5 | 1056 | |||
XM_017014805.1 | XP_016870294.1 | 4 | 818 | |||
XM_017014806.1 | XP_016870295.1 | 5 | 870 | |||
GenBank mRNA | AF070572.1 | - | 5 | 894 | ||
AF188239.1 | - | 7 | 1206 | |||
AK074844.1 | - | 5 | 1568 | |||
BC082765.1 | - | 6 | 1370 | |||
BC098265.1 | - | 4 | 788 | 40025067 | ||
BC098296.1 | - | 4 | 788 | 40025068 | ||
BC099733.1 | - | 4 | 788 | 40025069 |