Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 7, 8 | 749 | ||
XM_011518780.2 | XP_011517082.1 | 6, 7 | 686 | |||
GenBank mRNA | AF151054.1 | - | 7, 8 | 740 | ||
AF218421.1 | - | 7, 8 | 704 | |||
AK057004.1 | - | 7, 8 | 731 | |||
AK096971.1 | - | 1, 2 | 1279 | |||
AK097577.1 | - | 4, 5 | 1511 | |||
AL137549.1 | - | 5, 6 | 535 | |||
AY632862.1 | - | 4, 5 | 440 | |||
BC007664.2 | - | 7, 8 | 702 | 3140172 | ||
BC017570.1 | - | 7, 8 | 706 | 4109447 | ||
BC037173.1 | - | 2, 3 | 255 |