Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016530.2 | NP_057614.1 | 2 | 250 | ||
XM_011521669.2 | XP_011519971.1 | 2 | 243 | |||
XM_017022312.1 | XP_016877801.1 | 3 | 305 | |||
GenBank mRNA | AB038995.1 | - | 2 | 245 | ||
AK001111.1 | - | 2 | 173 | |||
AK095444.1 | - | 2 | 173 | |||
AK297598.1 | - | 2 | 173 | |||
AL833365.1 | - | 2 | 163 | |||
BC020654.1 | - | 2 | 162 | 4701429 | ||
BG721755.1 | - | 2 | 314 |