Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016541.2 | NP_057625.1 | 3 | 884 | ||
GenBank mRNA | AB030207.1 | - | 3 | 863 | ||
AY029486.1 | - | 3 | 850 | |||
BM931126.1 | - | 1 | 433 |