Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 9 | 967 | ||
NM_001042453.1 | NP_001035918.1 | 10 | 1213 | |||
NM_016542.3 | NP_057626.2 | 11 | 1444 | |||
XM_011531349.1 | XP_011529651.1 | 12 | 1417 | |||
XM_011531350.1 | XP_011529652.1 | 11 | 1490 | |||
GenBank mRNA | AB040057.2 | - | 11 | 1363 | ||
AF231012.1 | - | 11 | 1332 | |||
AF344882.1 | - | 10 | 1153 | |||
AF344883.1 | - | 9 | 967 | |||
AK000556.1 | - | 7 | 776 | |||
AK027589.1 | - | 6 | 661 | |||
AK074837.1 | - | 6 | 697 | |||
AK075107.1 | - | 10 | 1125 | |||
AK075150.1 | - | 10 | 1287 | |||
AK303583.1 | - | 12 | 1425 | |||
AK314356.1 | - | 11 | 1360 | |||
AK316137.1 | - | 12 | 1425 | |||
BC098315.1 | - | 11 | 1349 | 40026349 | ||
BC099843.1 | - | 11 | 1349 | 40026348 | ||
BC103503.1 | - | 11 | 1349 | 40026350 | ||
BT020099.1 | - | 10 | 1143 |