Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017419.2 | NP_059115.1 | 9 | 1318 | ||
XM_017008291.1 | XP_016863780.1 | 8 | 2222 | |||
GenBank mRNA | AJ252011.1 | - | 9 | 1318 |