Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282225.1 | NP_001269154.1 | 9, 10 | 1716 | ||
NM_001282226.1 | NP_001269155.1 | 9, 10 | 1683 | |||
NM_001282227.1 | NP_001269156.1 | 9, 10 | 1489 | |||
NM_001282228.1 | NP_001269157.1 | 9, 10 | 1591 | |||
NM_001282229.1 | NP_001269158.1 | 8, 9 | 1348 | |||
NM_177405.2 | NP_803124.1 | 6, 7 | 922 | |||
XM_006724080.3 | XP_006724143.1 | 8, 9 | 2953 | |||
XM_011546133.1 | XP_011544435.1 | 9, 10 | 1608 | |||
GenBank mRNA | AB209648.1 | - | 8, 9 | 2953 | ||
AF190746.1 | - | 8, 9 | 1652 | |||
AK027682.1 | - | 6, 7 | 743 | |||
AK074702.1 | - | 6, 7 | 794 | |||
AK130725.1 | - | 3, 4 | 436 | |||
AK292689.1 | - | 9, 10 | 1704 | |||
AK304818.1 | - | 9, 10 | 1483 | |||
AK314321.1 | - | 9, 10 | 1507 | |||
BC051755.1 | - | 8, 9 | 1487 | 5753141 | ||
CR456417.1 | - | 8, 9 | 1474 |