Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPXCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184771.1 | NP_001171700.1 | 3 | 883 | ||
NM_033048.5 | NP_149037.4 | 3 | 921 | |||
GenBank mRNA | AF284767.1 | - | 1 | 698 | ||
AK098646.1 | - | 3 | 858 | |||
AK313768.1 | - | 3 | 896 | |||
BC027614.2 | - | 3 | 891 | 4838752 |