Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138284.1 | NP_612141.1 | 3 | 1595 | ||
XM_005266421.3 | XP_005266478.1 | 3 | 2039 | |||
XM_005266422.2 | XP_005266479.1 | 2 | 1756 | |||
GenBank mRNA | AK123964.1 | - | 3 | 1678 | ||
AY078238.1 | - | 3 | 1595 | |||
AY359113.1 | - | 2 | 1487 | |||
BC036243.1 | - | 3 | 1598 | 5301608 |