Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024047.4 | NP_076952.1 | 3 | 719 | ||
NM_198038.2 | NP_932155.1 | 3 | 361 | |||
XM_011532031.2 | XP_011530333.1 | 3 | 722 | |||
XM_011532032.2 | XP_011530334.1 | 3 | 2898 | |||
GenBank mRNA | AF265341.1 | - | 3 | 360 | ||
AF273028.1 | - | 3 | 683 | |||
AK027295.1 | - | 3 | 709 | |||
AK074845.1 | - | 3 | 361 | |||
AK075408.1 | - | 3 | 639 | |||
AY026252.1 | - | 3 | 714 | |||
AY359123.1 | - | 3 | 413 | |||
BC000542.2 | - | 3 | 479 | 3163944 | ||
BI552154.1 | - | 359 | ||||
BM544951.1 | - | 3 | 731 |