Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039840.2 | NP_001034929.2 | 5, 6 | 760 | ||
XM_017029583.1 | XP_016885072.1 | 6, 7 | 441 | |||
GenBank mRNA | AK297768.1 | - | 5, 6 | 617 | ||
AL832095.1 | - | 5, 6 | 400 | |||
BC127728.1 | - | 5, 6 | 671 | 40127525 | ||
BC136691.1 | - | 5, 6 | 665 | 9020681 | ||
BC136692.1 | - | 5, 6 | 665 | 9020682 |