Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 5, 6 | 598 | ||
NM_023003.3 | NP_075379.2 | 5, 6 | 598 | |||
XM_011521679.2 | XP_011519981.1 | 5, 6 | 523 | |||
XM_011521680.2 | XP_011519982.1 | 5, 6 | 523 | |||
XM_017022320.1 | XP_016877809.1 | 5, 6 | 523 | |||
XM_017022321.1 | XP_016877810.1 | 5, 6 | 523 | |||
XM_017022322.1 | XP_016877811.1 | 5, 6 | 523 | |||
XM_017022323.1 | XP_016877812.1 | 5, 6 | 650 | |||
XM_017022324.1 | XP_016877813.1 | 5, 6 | 627 | |||
XM_017022325.1 | XP_016877814.1 | 5, 6 | 512 | |||
XM_017022326.1 | XP_016877815.1 | 5, 6 | 512 | |||
XM_017022327.1 | XP_016877816.1 | 5, 6 | 704 | |||
GenBank mRNA | AF255922.1 | - | 5, 6 | 546 | ||
AK223246.1 | - | 5, 6 | 543 | |||
AK292100.1 | - | 5, 6 | 523 | |||
AK295568.1 | - | 5, 6 | 487 | |||
DQ021909.1 | - | 5, 6 | 489 |