Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281734.1 | NP_001268663.1 | 7 | 2216 | ||
NM_001281735.1 | NP_001268664.1 | 4 | 352 | |||
NM_021260.3 | NP_067083.1 | 7 | 2216 | |||
NM_178441.2 | NP_848535.1 | 4 | 357 | |||
XM_017021373.1 | XP_016876862.1 | 5 | 881 | |||
GenBank mRNA | AB046809.1 | - | 7 | 2041 | ||
AF251025.2 | - | 6 | 1921 | |||
AF311602.1 | - | 4 | 357 | |||
AF318319.1 | - | 4 | 385 | |||
AJ310569.1 | - | 7 | 2067 | |||
AK027399.1 | - | 7 | 2110 | |||
AK098542.1 | - | 4 | 340 | |||
BC014902.1 | - | 7 | 2135 | |||
BC053520.1 | - | 7 | 2144 | 5493027 |