Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281734.1 | NP_001268663.1 | 11, 12 | 2783 | ||
NM_001281735.1 | NP_001268664.1 | 8, 9 | 961 | |||
NM_021260.3 | NP_067083.1 | 11, 12 | 2825 | |||
NM_178441.2 | NP_848535.1 | 8, 9 | 966 | |||
XM_017021373.1 | XP_016876862.1 | 9, 10 | 1490 | |||
GenBank mRNA | AB046809.1 | - | 11, 12 | 2650 | ||
AF251025.2 | - | 10, 11 | 2530 | |||
AF311602.1 | - | 8, 9 | 966 | |||
AF318319.1 | - | 8, 9 | 994 | |||
AJ310569.1 | - | 11, 12 | 2676 | |||
AK027399.1 | - | 11, 12 | 2719 | |||
AK098542.1 | - | 8, 9 | 949 | |||
BC014902.1 | - | 11, 12 | 2702 | |||
BC053520.1 | - | 11, 12 | 2753 | 5493027 |