Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138316.3 | NP_612189.2 | 3 | 703 | ||
NM_148977.2 | NP_683878.1 | 3 | 1481 | |||
NM_148978.2 | NP_683879.1 | 3 | 703 | |||
XM_017016333.1 | XP_016871822.1 | 4 | 1449 | |||
XM_017016334.1 | XP_016871823.1 | 4 | 1411 | |||
XM_017016335.1 | XP_016871824.1 | 4 | 2239 | |||
XM_017016336.1 | XP_016871825.1 | 3 | 628 | |||
XM_017016337.1 | XP_016871826.1 | 4 | 2145 | |||
GenBank mRNA | AF355198.1 | - | 3 | 687 | ||
AK125428.1 | - | 4 | 1060 | |||
AK127777.1 | - | 2 | 402 | |||
AY027661.1 | - | 3 | 1481 | |||
AY027662.1 | - | 3 | 703 | |||
AY027663.1 | - | 4 | 797 |