Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016930.2 | NP_058626.1 | 10 | 985 | ||
XM_011513480.1 | XP_011511782.1 | 11 | 1057 | |||
XM_011513483.1 | XP_011511785.1 | 10 | 958 | |||
XM_011513485.2 | XP_011511787.1 | 10 | 912 | |||
XM_017008297.1 | XP_016863786.1 | 11 | 1638 | |||
GenBank mRNA | AB028741.1 | - | 10 | 904 | ||
AK310026.1 | - | 10 | 947 | |||
AL832867.1 | - | 4 | 251 | |||
AY453396.1 | - | 10 | 901 | |||
BC014613.2 | - | 10 | 907 | 2960768 | ||
BT007150.1 | - | 10 | 901 | |||
BT020135.1 | - | 10 | 901 | |||
BX647083.1 | - | 3 | 1590 |