Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022338.3 | NP_071733.1 | 4 | 704 | ||
XM_005274053.3 | XP_005274110.1 | 4 | 879 | |||
XM_011545103.2 | XP_011543405.1 | 2 | 299 | |||
GenBank mRNA | AF264781.1 | - | 4 | 664 | ||
AF370372.1 | - | 3 | 572 | |||
AY358754.1 | - | 2 | 266 | |||
BC011765.2 | - | 3 | 553 | 3614861 | ||
BG701401.1 | - | 4 | 710 |