Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 4 | 646 | ||
NM_001276290.1 | NP_001263219.1 | 2 | 343 | |||
NM_001278433.1 | NP_001265362.1 | 3 | 516 | |||
NM_002734.4 | NP_002725.1 | 3 | 513 | |||
NM_212471.2 | NP_997636.1 | 3 | 706 | |||
NM_212472.2 | NP_997637.1 | 3 | 544 | |||
XM_011524983.2 | XP_011523285.1 | 3 | 609 | |||
XM_011524984.2 | XP_011523286.1 | 4 | 547 | |||
XM_011524985.2 | XP_011523287.1 | 4 | 841 | |||
GenBank mRNA | A12295.1 | - | 3 | 439 | ||
AK097580.1 | - | 2 | 916 | |||
AK124586.1 | - | 3 | 432 | |||
AK312307.1 | - | 3 | 442 | |||
AL535889.3 | - | 3 | 484 | |||
BC036285.1 | - | 3 | 465 | 4340015 | ||
BC093042.1 | - | 3 | 454 | 6514677 | ||
BM928493.1 | - | 3 | 511 | |||
BU167718.1 | - | 3 | 553 | |||
BU188258.1 | - | 3 | 506 | |||
CR749311.1 | - | 4 | 575 | |||
DA108801.1 | - | 3 | 518 | |||
DA219203.1 | - | 3 | 513 | |||
M18468.1 | - | 3 | 440 | |||
M33336.1 | - | 3 | 424 |