Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243270.1 | NP_001230199.1 | 16 | 2397 | ||
NM_001243271.1 | NP_001230200.1 | 17 | 2536 | |||
NM_014361.3 | NP_055176.1 | 17 | 2536 | |||
NM_175566.2 | NP_780775.1 | 14 | 1880 | |||
XM_011542871.1 | XP_011541173.1 | 16 | 2314 | |||
XM_017017926.1 | XP_016873415.1 | 18 | 2704 | |||
GenBank mRNA | AB013802.2 | - | 15 | 2111 | ||
AB013803.2 | - | 14 | 1889 | |||
AK294692.1 | - | 3 | 280 | |||
BC039255.1 | - | 17 | 2371 | 5273769 | ||
BC130619.1 | - | 16 | 2183 | 40146650 | ||
BC144185.1 | - | 16 | 2183 | 9052705 | ||
BC144186.1 | - | 17 | 2322 | 9052706 |