Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A4GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318038.1 | NP_001304967.1 | 3 | 868 | ||
NM_017436.5 | NP_059132.1 | 3 | 845 | |||
XM_005261644.2 | XP_005261701.1 | 3 | 1150 | |||
XM_005261646.4 | XP_005261703.1 | 2 | 706 | |||
XM_005261647.2 | XP_005261704.1 | 3 | 680 | |||
XM_005261648.3 | XP_005261705.1 | 3 | 763 | |||
XM_006724265.3 | XP_006724328.1 | 3 | 1091 | |||
XM_006724266.3 | XP_006724329.1 | 3 | 825 | |||
XM_011530233.2 | XP_011528535.1 | 2 | 1032 | |||
XM_017028831.1 | XP_016884320.1 | 2 | 962 | |||
GenBank mRNA | AB037883.1 | - | 2 | 677 | ||
AJ245581.1 | - | 1 | 544 | |||
AK074388.1 | - | 1 | 444 | |||
AK312927.1 | - | 3 | 849 | |||
BC017068.2 | - | 3 | 764 | 3913851 | ||
BC055286.1 | - | 2 | 1034 | 6161307 | ||
KU178649.1 | - | 1 | 544 |