Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320602.1 | NP_001307531.1 | 5 | 546 | ||
NM_144770.4 | NP_658983.3 | 5 | 525 | |||
NR_135308.1 | 4 | 362 | ||||
NR_135309.1 | 4 | 383 | ||||
NR_135310.1 | 5 | 535 | ||||
NR_135311.1 | 5 | 556 | ||||
XM_005260997.4 | XP_005261054.1 | 5 | 569 | |||
XM_017028384.1 | XP_016883873.1 | 4 | 284 | |||
XM_017028385.1 | XP_016883874.1 | 4 | 3712 | |||
XM_017028386.1 | XP_016883875.1 | 5 | 680 | |||
XM_017028387.1 | XP_016883876.1 | 5 | 687 | |||
GenBank mRNA | AF519623.1 | - | 4 | 326 | ||
AK091331.1 | - | 5 | 507 | |||
AY077695.1 | - | 5 | 456 | |||
BC030196.1 | - | 5 | 477 | 5199053 | ||
CB153254.1 | - | 5 | 535 | |||
CN430785.1 | - | 5 | 542 |