Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017442.3 | NP_059138.1 | 2 | 2231 | ||
GenBank mRNA | AB045180.1 | - | 2 | 1703 | ||
AB445673.1 | - | 1 | 1597 | |||
AF259262.1 | - | 2 | 2231 | |||
AF259263.1 | - | 1 | 1473 | |||
AY359085.1 | - | 1 | 1607 | |||
BC032713.1 | - | 2 | 1693 | 5495717 |