Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 6 | 808 | ||
NM_020640.3 | NP_065691.2 | 6 | 790 | |||
XM_011512912.1 | XP_011511214.1 | 6 | 759 | |||
XM_011512914.1 | XP_011511216.1 | 6 | 798 | |||
XM_011512915.2 | XP_011511217.1 | 7 | 1320 | |||
XM_011512916.2 | XP_011511218.1 | 6 | 1248 | |||
XM_017006633.1 | XP_016862122.1 | 5 | 675 | |||
XM_017006634.1 | XP_016862123.1 | 6 | 764 | |||
XM_017006635.1 | XP_016862124.1 | 7 | 1320 | |||
GenBank mRNA | AF292100.2 | - | 6 | 707 | ||
AF456425.1 | - | 6 | 693 | |||
AF456426.1 | - | 6 | 693 | |||
AK025764.1 | - | 4 | 353 | |||
AK056335.1 | - | 6 | 723 | |||
AK297328.1 | - | 7 | 607 | |||
AK314480.1 | - | 6 | 707 | |||
BC009478.2 | - | 6 | 712 | 3939758 | ||
BC013163.1 | - | 5 | 663 |