Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321073.1 | NP_001308002.1 | 7, 8 | 910 | ||
XM_011546094.1 | XP_011544396.1 | 7, 8 | 802 | |||
XM_011546095.2 | XP_011544397.1 | 7, 8 | 944 | |||
XM_011546097.2 | XP_011544399.1 | 8, 9 | 971 | |||
XM_011546098.1 | XP_011544400.1 | 7, 8 | 1309 | |||
XM_011546099.1 | XP_011544401.1 | 7, 8 | 802 | |||
XM_017028209.1 | XP_016883698.1 | 5, 6 | 1007 | |||
XM_017028210.1 | XP_016883699.1 | 5, 6 | 1007 | |||
XM_017028211.1 | XP_016883700.1 | 8, 9 | 1307 | |||
XM_017028212.1 | XP_016883701.1 | 9, 10 | 1363 | |||
XM_017028213.1 | XP_016883702.1 | 7, 8 | 802 | |||
XM_017028214.1 | XP_016883703.1 | 8, 9 | 1304 | |||
XM_017028215.1 | XP_016883704.1 | 8, 9 | 919 | |||
XM_017028216.1 | XP_016883705.1 | 7, 8 | 1309 | |||
XM_017028217.1 | XP_016883706.1 | 7, 8 | 802 | |||
XM_017028218.1 | XP_016883707.1 | 8, 9 | 971 | |||
GenBank mRNA | BC007257.1 | - | 7, 8 | 880 | 3028099 |