Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021161.4 | NP_066984.1 | 5 | 1212 | ||
NM_138317.2 | NP_612190.1 | 5 | 1221 | |||
NM_138318.2 | NP_612191.1 | 5 | 892 | |||
XM_011536840.2 | XP_011535142.1 | 4 | 597 | |||
GenBank mRNA | AF279890.1 | - | 5 | 1227 | ||
AF385399.1 | - | 5 | 893 | |||
AF385400.1 | - | 5 | 975 | |||
AK290518.1 | - | 5 | 1160 | |||
AK313499.1 | - | 5 | 1221 | |||
AK315263.1 | - | 5 | 880 | |||
BC075021.2 | - | 5 | 811 | 30915621 | ||
BC075022.2 | - | 5 | 808 | 30915410 | ||
EU978938.1 | - | 5 | 1041 | |||
EU978939.1 | - | 5 | 894 | |||
EU978940.1 | - | 5 | 798 | |||
EU978941.1 | - | 5 | 798 |