Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285878.1 | NP_001272807.1 | 1 | 511 | ||
NM_001285879.1 | NP_001272808.1 | 1 | 511 | |||
NM_005194.3 | NP_005185.2 | 1 | 511 | |||
GenBank mRNA | AF289608.1 | - | 1 | 252 | ||
AK291536.1 | - | 1 | 264 | |||
BC007538.1 | - | 1 | 240 | 3028673 | ||
BC021931.1 | - | 1 | 255 | 4870789 | ||
KJ161450.1 | - | 1 | 35 | |||
KJ161451.1 | - | 1 | 35 | |||
KJ161452.1 | - | 1 | 35 | |||
KJ161453.1 | - | 1 | 35 | |||
KJ161454.1 | - | 1 | 35 | |||
KJ161455.1 | - | 1 | 35 | |||
KJ161456.1 | - | 1 | 35 | |||
KJ161457.1 | - | 1 | 35 | |||
KJ161458.1 | - | 1 | 35 | |||
KJ161459.1 | - | 1 | 35 | |||
KJ161460.1 | - | 1 | 35 | |||
KJ161461.1 | - | 1 | 35 | |||
KJ161462.1 | - | 1 | 35 | |||
KJ161463.1 | - | 1 | 35 | |||
KJ161464.1 | - | 1 | 35 | |||
KJ161465.1 | - | 1 | 35 | |||
KJ161466.1 | - | 1 | 35 | |||
KJ161467.1 | - | 1 | 35 | |||
KJ161468.1 | - | 1 | 35 | |||
KJ161469.1 | - | 1 | 35 | |||
KJ161470.1 | - | 1 | 35 | |||
KJ161471.1 | - | 1 | 35 | |||
KJ161472.1 | - | 1 | 35 | |||
KJ161473.1 | - | 1 | 35 | |||
KJ161474.1 | - | 1 | 35 | |||
KJ161475.1 | - | 1 | 35 | |||
KJ161476.1 | - | 1 | 35 | |||
KJ161477.1 | - | 1 | 35 | |||
KJ161478.1 | - | 1 | 35 | |||
KJ161479.1 | - | 1 | 35 | |||
KJ161480.1 | - | 1 | 35 |