Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287813.2 | NP_001274742.1 | 17 | 1605 | ||
NM_001287814.2 | NP_001274743.1 | 15 | 1757 | |||
NM_001321773.1 | NP_001308702.1 | 15 | 1757 | |||
NM_001321775.1 | NP_001308704.1 | 17 | 1605 | |||
NM_001321776.1 | NP_001308705.1 | 16 | 1832 | |||
NM_001321777.1 | NP_001308706.1 | 15 | 1757 | |||
NM_001321778.1 | NP_001308707.1 | 16 | 1563 | |||
NM_018967.4 | NP_061840.1 | 16 | 1832 | |||
NR_135794.1 | 16 | 1812 | ||||
NR_135795.1 | 16 | 1827 | ||||
NR_135796.1 | 16 | 1858 | ||||
NR_135797.1 | 18 | 2002 | ||||
NR_135798.1 | 15 | 1752 | ||||
XM_011517548.2 | XP_011515850.1 | 15 | 1942 | |||
XM_017013579.1 | XP_016869068.1 | 18 | 4761 | |||
XM_017013580.1 | XP_016869069.1 | 16 | 1355 | |||
GenBank mRNA | AJ003030.1 | - | 15 | 1393 | ||
AK314084.1 | - | 17 | 1542 | |||
AL161971.1 | - | 16 | 1727 | |||
BC075072.2 | - | 14 | 1197 | 30915592 | ||
BC104829.1 | - | 15 | 1279 | 8143832 | ||
BC143695.1 | - | 15 | 1305 | |||
BG194056.1 | - | 1 | 170 |