Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018968.3 | NP_061841.2 | 8 | 695 | ||
XM_011510366.2 | XP_011508668.1 | 6 | 732 | |||
XM_017004361.1 | XP_016859850.1 | 8 | 657 | |||
XM_017004362.1 | XP_016859851.1 | 8 | 663 | |||
XM_017004363.1 | XP_016859852.1 | 8 | 844 | |||
XM_017004364.1 | XP_016859853.1 | 5 | 3985 | |||
XM_017004365.1 | XP_016859854.1 | 8 | 658 | |||
XM_017004366.1 | XP_016859855.1 | 8 | 658 | |||
XM_017004367.1 | XP_016859856.1 | 9 | 733 | |||
XM_017004368.1 | XP_016859857.1 | 8 | 658 | |||
XM_017004369.1 | XP_016859858.1 | 8 | 658 | |||
XM_017004370.1 | XP_016859859.1 | 8 | 658 | |||
XM_017004371.1 | XP_016859860.1 | 8 | 659 | |||
XM_017004372.1 | XP_016859861.1 | 8 | 659 | |||
XM_017004373.1 | XP_016859862.1 | 3 | 1299 | |||
XM_017004374.1 | XP_016859863.1 | 8 | 664 | |||
GenBank mRNA | AJ003029.1 | - | 8 | 697 |